Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RMDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304802.1 | NP_001291731.1 | 6 | 1123 | ||
NM_001323894.1 | NP_001310823.1 | 6 | 1037 | |||
NM_001323895.1 | NP_001310824.1 | 6 | 1254 | |||
NM_001323896.1 | NP_001310825.1 | 6 | 1115 | |||
NM_001323897.1 | NP_001310826.1 | 6 | 1231 | |||
NM_018145.2 | NP_060615.1 | 6 | 1153 | |||
XM_011521755.2 | XP_011520057.1 | 3 | 1399 | |||
GenBank mRNA | AB000782.1 | - | 5 | 892 | ||
AK001441.1 | - | 6 | 1077 | |||
AK025963.1 | - | 3 | 466 | |||
AK092058.1 | - | 6 | 1036 | |||
AK097286.1 | - | 7 | 965 | |||
AK123192.1 | - | 7 | 1031 | |||
AK123282.1 | - | 5 | 1960 | |||
AY358793.1 | - | 5 | 893 | |||
BC008970.2 | - | 2 | 350 | |||
BC063844.1 | - | 6 | 1041 | 6527521 |