Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SPATA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258372.1 | NP_001245301.1 | 14, 15 | 2234 | ||
NM_001258373.1 | NP_001245302.1 | 15, 16 | 2337 | |||
NM_022827.3 | NP_073738.2 | 15, 16 | 2282 | |||
GenBank mRNA | AF086245.1 | - | 1, 2 | 129 | ||
AK025000.1 | - | 14, 15 | 2131 | |||
AK025622.1 | - | 10, 11 | 1523 | |||
AK092731.1 | - | 15, 16 | 2267 | |||
AK125807.1 | - | 16, 17 | 3104 | |||
AK225189.1 | - | 14, 15 | 2131 | |||
AK291805.1 | - | 15, 16 | 2225 | |||
BC017468.1 | - | 4, 5 | 726 | |||
BC025255.1 | - | 15, 16 | 2170 | 5019010 | ||
BC065526.1 | - | 15, 16 | 2224 | 6149633 | ||
BC111029.1 | - | 14, 15 | 2139 | 5245538 | ||
CR936769.1 | - | 12, 13 | 2761 | |||
DQ238597.1 | - | 14, 15 | 2232 | |||
GQ472220.1 | - | 14, 15 | 2139 |