Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MFSD11 SIRNA Assays ›
Gene Symbol:
MFSD11
Gene Name:
major facilitator superfamily domain containing 11
Gene Aliases:
ET
Chromosome Location:
Chr.17: 76736565 - 76803805 on Build GRCh38
UniGene ID:
Hs.732019
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
140788
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MFSD11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242532.1
NP_001229461.1
11
1759
NM_001242533.1
NP_001229462.1
12
1643
NM_001242534.1
NP_001229463.1
12
1505
NM_001242535.1
NP_001229464.1
12
1454
NM_001242536.1
NP_001229465.1
10
1487
NM_001242537.1
NP_001229466.1
10
1298
NM_024311.3
NP_077287.1
12
1361
XM_011525236.2
XP_011523538.1
11
1023
XM_011525237.2
XP_011523539.2
12
1466
XM_011525238.2
XP_011523540.2
12
1066
XM_011525239.2
XP_011523541.2
12
1036
XM_011525240.2
XP_011523542.2
12
1062
XM_011525241.2
XP_011523543.2
12
1072
XM_011525242.1
XP_011523544.1
11
1023
XM_011525244.1
XP_011523546.1
11
1023
XM_011525247.2
XP_011523549.2
12
1506
XM_017025065.1
XP_016880554.1
12
1094
XM_017025066.1
XP_016880555.1
12
1036
XM_017025067.1
XP_016880556.1
12
1094
XM_017025068.1
XP_016880557.1
12
1506
XM_017025069.1
XP_016880558.1
10
1194
XM_017025070.1
XP_016880559.1
10
1194
XM_017025071.1
XP_016880560.1
4
383
GenBank mRNA
AF015184.1
-
12
1073
AF015185.1
-
10
1104
AF015186.1
-
12
1217
AK000233.1
-
11
1141
AK025849.1
-
11
896
AK308058.1
-
10
942
BC002753.2
-
12
1161
3629029
BC062563.1
-
12
1254
5140496
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-vvmmd:80/10.239.160.160:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0