Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NLRC5 SIRNA Assays ›
          Gene Symbol:
          NLRC5
          Gene Name:
          NLR family CARD domain containing 5
          Gene Aliases:
          CLR16.1, NOD27, NOD4
          Chromosome Location:
          Chr.16: 56989547 - 57083524 on Build GRCh38
          UniGene ID:
          Hs.528836
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 141059

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NLRC5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_032206.4 NP_115582.4 5 1653
          XM_005256193.2 XP_005256250.1 6 1714
          XM_005256194.3 XP_005256251.1 6 1925
          XM_005256195.2 XP_005256252.1 6 1714
          XM_005256196.2 XP_005256253.1 6 1714
          XM_005256197.2 XP_005256254.1 6 1714
          XM_005256199.3 XP_005256256.1 6 1714
          XM_005256201.3 XP_005256258.1 6 1714
          XM_006721297.2 XP_006721360.1 6 1702
          XM_006721298.2 XP_006721361.1 6 1714
          XM_006721300.2 XP_006721363.1 6 1714
          XM_011523373.2 XP_011521675.1 6 1861
          XM_011523375.2 XP_011521677.1 6 1714
          XM_011523376.2 XP_011521678.1 1 22
          XM_017023766.1 XP_016879255.1 6 1714
          XM_017023767.1 XP_016879256.1 6 1702
          XM_017023768.1 XP_016879257.1 6 1714
          XM_017023769.1 XP_016879258.1 6 1702
          XM_017023770.1 XP_016879259.1 6 1714
          XM_017023771.1 XP_016879260.1 6 1714
          GenBank mRNA AF389420.1 - 4 1526
          AK025362.1 - 1 785
          AK074182.1 - 5 1666
          AK090439.1 - 1 708
          AK097030.1 - 6 1703
          BX640842.1 - 2 188
          EF452236.1 - 4 1526

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of NF-kappaB transcription factor activity
          negative regulation of type I interferon production
          regulation of kinase activity
          innate immune response
          positive regulation of MHC class I biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          defense response to virus
          positive regulation of interferon-gamma-mediated signaling pathway
          negative regulation of type I interferon-mediated signaling pathway
          positive regulation of type I interferon-mediated signaling pathway
          RNA polymerase II core promoter sequence-specific DNA binding
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline