Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FRMD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286490.1 | NP_001273419.1 | 16 | 1721 | ||
NM_001286491.1 | NP_001273420.1 | 15 | 2284 | |||
NM_001322950.1 | NP_001309879.1 | 15 | 1786 | |||
NM_032892.4 | NP_116281.2 | 14 | 1965 | |||
NR_104455.1 | 15 | 2281 | ||||
GenBank mRNA | AK090572.1 | - | 14 | 1897 | ||
AK123018.1 | - | 16 | 1721 | |||
AK290013.1 | - | 3 | 759 | |||
BC043486.2 | - | 15 | 2031 | |||
BC053647.1 | - | 14 | 1902 | 5578205 |