Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SARNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033082.3 | NP_149073.1 | 8, 9 | 490 | ||
NR_026722.1 | 7, 8 | 443 | ||||
NR_026723.1 | 8, 9 | 490 | ||||
GenBank mRNA | AF161434.1 | - | 8, 9 | 459 | ||
AF486281.1 | - | 8, 9 | 435 | |||
AK290508.1 | - | 8, 9 | 458 | |||
BC007099.1 | - | 8, 9 | 456 | 4273903 | ||
BC093051.1 | - | 7, 8 | 409 | 30382893 | ||
BI520421.1 | - | 8, 9 | 490 |