Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBFOX1 SIRNA Assays ›
Gene Symbol:
RBFOX1
Gene Name:
RNA binding protein, fox-1 homolog 1
Gene Aliases:
2BP1, A2BP1, FOX-1, FOX1, HRNBP1
Chromosome Location:
Chr.16: 5239749 - 7713343 on Build GRCh38
UniGene ID:
Hs.459842
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
141407
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBFOX1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142333.1
NP_001135805.1
7, 8
1452
NM_001142334.1
NP_001135806.1
5, 6
684
NM_001308117.1
NP_001295046.1
7, 8
789
NM_018723.3
NP_061193.2
7, 8
1452
NM_145891.2
NP_665898.1
4, 5
776
NM_145892.2
NP_665899.1
4, 5
776
NM_145893.2
NP_665900.1
4, 5
776
XM_005255386.3
XP_005255443.1
4, 5
702
XM_005255387.3
XP_005255444.1
4, 5
681
XM_005255388.4
XP_005255445.1
4, 5
737
XM_005255390.3
XP_005255447.1
7, 8
1574
XM_005255391.3
XP_005255448.1
6, 7
584
XM_005255394.4
XP_005255451.1
4, 5
681
XM_011522546.2
XP_011520848.1
4, 5
681
XM_011522547.2
XP_011520849.1
4, 5
681
XM_011522548.2
XP_011520850.1
4, 5
681
XM_017023318.1
XP_016878807.1
8, 9
1079
XM_017023319.1
XP_016878808.1
10, 11
1075
XM_017023320.1
XP_016878809.1
7, 8
1483
XM_017023321.1
XP_016878810.1
6, 7
2545
XM_017023322.1
XP_016878811.1
6, 7
2545
XM_017023323.1
XP_016878812.1
4, 5
702
XM_017023324.1
XP_016878813.1
7, 8
1483
XM_017023325.1
XP_016878814.1
5, 6
1081
XM_017023326.1
XP_016878815.1
7, 8
1574
XM_017023327.1
XP_016878816.1
4, 5
702
XM_017023328.1
XP_016878817.1
7, 8
1483
XM_017023329.1
XP_016878818.1
7, 8
1483
XM_017023330.1
XP_016878819.1
6, 7
2022
XM_017023331.1
XP_016878820.1
4, 5
2488
XM_017023332.1
XP_016878821.1
4, 5
702
XM_017023333.1
XP_016878822.1
6, 7
584
XM_017023334.1
XP_016878823.1
4, 5
702
XM_017023335.1
XP_016878824.1
7, 8
1574
XM_017023336.1
XP_016878825.1
6, 7
584
XM_017023337.1
XP_016878826.1
4, 5
702
XM_017023338.1
XP_016878827.1
6, 7
584
XM_017023339.1
XP_016878828.1
4, 5
728
XM_017023341.1
XP_016878830.1
7, 8
1574
XM_017023342.1
XP_016878831.1
6, 7
584
XM_017023343.1
XP_016878832.1
4, 5
731
GenBank mRNA
AB208826.1
-
1486
AF094849.1
-
4, 5
777
AF107203.1
-
7, 8
1450
AF109106.1
-
4, 5
777
AF229057.1
-
4, 5
779
AF448859.1
-
7, 8
1450
AK001027.1
-
5, 6
684
AK293951.1
-
7, 8
789
AL713700.1
-
6, 7
1299
BC113691.1
-
4, 5
570
8322743
BC143816.1
-
4, 5
570
KJ534926.1
-
4, 5
524
KJ535000.1
-
4, 5
524
KJ535001.1
-
4, 5
524
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
RNA splicing
RNA transport
nucleotide binding
RNA binding
mRNA binding
protein binding
protein C-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-sskjb:80/100.66.75.127:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0