Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATXN2L SIRNA Assays ›
Gene Symbol:
ATXN2L
Gene Name:
ataxin 2 like
Gene Aliases:
A2D, A2LG, A2LP, A2RP
Chromosome Location:
Chr.16: 28823048 - 28837237 on Build GRCh38
UniGene ID:
Hs.460499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
141410
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATXN2L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308230.1
NP_001295159.1
7
1003
NM_007245.3
NP_009176.2
7
1003
NM_017492.3
NP_059867.3
7
1003
NM_145714.2
NP_663760.1
7
1003
NM_148414.2
NP_680780.1
7
1003
NM_148415.2
NP_680781.1
7
1003
NM_148416.2
NP_680782.1
7
1003
XM_005255061.1
XP_005255118.1
7
1003
XM_005255062.1
XP_005255119.1
7
1003
XM_005255063.3
XP_005255120.1
7
1003
XM_005255064.1
XP_005255121.1
7
1003
XM_005255065.1
XP_005255122.1
7
1003
XM_005255066.1
XP_005255123.1
7
1003
XM_005255067.1
XP_005255124.1
7
1003
XM_005255068.1
XP_005255125.1
7
1003
XM_005255069.1
XP_005255126.1
7
1003
XM_005255070.1
XP_005255127.1
7
1003
XM_005255071.1
XP_005255128.1
7
1003
XM_005255074.1
XP_005255131.1
7
1003
XM_005255075.1
XP_005255132.1
7
1003
XM_005255076.1
XP_005255133.1
7
1003
XM_005255077.1
XP_005255134.1
7
886
XM_006721007.1
XP_006721070.1
7
1003
XM_006721008.1
XP_006721071.1
7
1003
XM_006721009.1
XP_006721072.1
7
1003
XM_006721010.2
XP_006721073.1
7
1003
XM_006721011.1
XP_006721074.1
7
1003
XM_006721012.3
XP_006721075.1
7
1003
XM_011545719.2
XP_011544021.1
7
1003
XM_011545721.2
XP_011544023.1
7
1003
XM_011545722.1
XP_011544024.1
7
703
XM_017022891.1
XP_016878380.1
7
886
XM_017022892.1
XP_016878381.1
7
703
GenBank mRNA
AF034373.1
-
9
1105
AJ317970.1
-
7
959
AJ317971.2
-
7
959
AJ317972.2
-
7
959
AJ317973.2
-
7
959
AJ317974.2
-
7
959
AK289981.1
-
7
1003
AY188334.1
-
9
1105
AY188335.1
-
9
1105
AY188336.1
-
9
1105
AY188337.1
-
9
1105
AY188338.1
-
9
1105
BC068012.1
-
8
861
BC082760.1
-
7
799
5812230
BC136584.1
-
7
956
9020574
U70671.1
-
7
633
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytoplasmic mRNA processing body assembly
stress granule assembly
cell-cell adhesion
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0