Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152464.2 | NP_689677.1 | 6 | 1149 | ||
GenBank mRNA | AK311363.1 | - | 1031 | |||
BC033113.1 | - | 6 | 1066 | 5429060 |