Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KRT26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181539.4 | NP_853517.2 | 3 | 656 | ||
GenBank mRNA | AJ564205.2 | - | 3 | 656 | ||
BC132951.1 | - | 3 | 647 | 40146973 | ||
BC136896.1 | - | 3 | 647 | 9020887 |