Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FDXR SIRNA Assays ›
          Gene Symbol:
          FDXR
          Gene Name:
          ferredoxin reductase
          Gene Aliases:
          ADXR
          Chromosome Location:
          Chr.17: 74862497 - 74873031 on Build GRCh38
          UniGene ID:
          Hs.69745
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 14192

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FDXR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001258012.3 NP_001244941.2 7, 8 941
          NM_001258013.3 NP_001244942.2 8, 9 905
          NM_001258014.3 NP_001244943.2 7, 8 788
          NM_001258015.2 NP_001244944.1 6, 7 692
          NM_001258016.3 NP_001244945.2 6, 7 757
          NM_004110.5 NP_004101.3 7, 8 830
          NM_024417.4 NP_077728.3 7, 8 812
          NR_047576.2 7, 8 912
          XM_006721772.1 XP_006721835.1 7, 8 760
          XM_011524527.1 XP_011522829.1 7, 8 778
          XM_011524528.1 XP_011522830.1 7, 8 867
          XM_011524529.1 XP_011522831.1 7, 8 870
          XM_011524531.2 XP_011522833.1 6, 7 763
          XM_011524532.2 XP_011522834.1 6, 7 770
          XM_011524533.2 XP_011522835.1 6, 7 738
          GenBank mRNA AK094120.1 - 8, 9 1010
          AK098648.1 - 7, 8 749
          AK223530.1 - 7, 8 742
          AK293208.1 - 8, 9 850
          AK293210.1 - 7, 8 849
          AK295307.1 - 7, 8 941
          AK298908.1 - 6, 7 635
          AK301779.1 - 7, 8 725
          AK301977.1 - 6, 7 757
          BC002960.1 - 7, 8 751 3537008
          BC018911.2 - 7, 8 758
          BC034763.1 - 7, 8 1413
          BC063493.1 - 7, 8 750 5404814
          BT006684.1 - 7, 8 725
          J03826.1 - 7, 8 763

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_024153, BC088844, D63761.
          This siRNA also targets the following transcripts in Mouse: NM_007997, AK134068, D49920, BC071268, BC057146.

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          generation of precursor metabolites and energy
          steroid biosynthetic process
          C21-steroid hormone biosynthetic process
          cholesterol metabolic process
          sterol metabolic process
          oxidation-reduction process
          ferredoxin-NADP+ reductase activity
          protein binding
          NADPH-adrenodoxin reductase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5btgp:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0