Search Thermo Fisher Scientific
Gene Symbol: LRRC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194287.2 | NP_919263.2 | 11, 12 | 1375 | ||
XM_011536479.1 | XP_011534781.1 | 11, 12 | 1743 | |||
XM_011536483.1 | XP_011534785.1 | 11, 12 | 1743 | |||
XM_017021032.1 | XP_016876521.1 | 11, 12 | 1641 | |||
XM_017021033.1 | XP_016876522.1 | 11, 12 | 1722 | |||
XM_017021034.1 | XP_016876523.1 | 11, 12 | 1387 | |||
XM_017021035.1 | XP_016876524.1 | 11, 12 | 1538 | |||
XM_017021036.1 | XP_016876525.1 | 9, 10 | 1611 | |||
XM_017021037.1 | XP_016876526.1 | 11, 12 | 2445 | |||
XM_017021038.1 | XP_016876527.1 | 11, 12 | 1279 | |||
XM_017021039.1 | XP_016876528.1 | 10, 11 | 1201 | |||
XM_017021040.1 | XP_016876529.1 | 11, 12 | 1200 | |||
XM_017021041.1 | XP_016876530.1 | 6, 7 | 699 | |||
GenBank mRNA | AK058106.1 | - | 10, 11 | 1288 | ||
AK125722.1 | - | 12, 13 | 1528 | |||
BC121159.2 | - | 11, 12 | 1223 | 40121413 | ||
BC121160.2 | - | 11, 12 | 1223 | 40121414 |