Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LARP4 SIRNA Assays ›
Gene Symbol:
LARP4
Gene Name:
La ribonucleoprotein domain family member 4
Gene Aliases:
PP13296
Chromosome Location:
Chr.12: 50400809 - 50480005 on Build GRCh38
UniGene ID:
Hs.26613
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
142015
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LARP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170803.1
NP_001164274.1
11
1270
NM_001170808.1
NP_001164279.1
12
1483
NM_052879.4
NP_443111.4
13
1696
NM_199188.2
NP_954658.2
13
1693
NM_199190.2
NP_954660.1
12
1483
XM_005268613.2
XP_005268670.1
14
1745
XM_011537828.2
XP_011536130.1
14
1800
XM_011537829.2
XP_011536131.1
14
1781
XM_011537830.2
XP_011536132.1
13
1628
XM_011537831.2
XP_011536133.1
13
1692
XM_011537833.2
XP_011536135.1
13
1587
XM_011537834.2
XP_011536136.1
13
1588
XM_011537835.2
XP_011536137.1
13
1568
XM_011537836.2
XP_011536138.1
13
1569
XM_011537838.2
XP_011536140.1
13
1540
XM_011537839.2
XP_011536141.1
14
1684
XM_011537840.2
XP_011536142.1
14
1737
XM_011537841.2
XP_011536143.1
13
1590
XM_011537843.2
XP_011536145.1
12
1356
XM_011537844.2
XP_011536146.1
14
1801
XM_017018750.1
XP_016874239.1
12
1409
XM_017018751.1
XP_016874240.1
14
1561
XM_017018752.1
XP_016874241.1
13
1544
XM_017018753.1
XP_016874242.1
13
1577
XM_017018754.1
XP_016874243.1
14
1727
XM_017018755.1
XP_016874244.1
12
1394
XM_017018756.1
XP_016874245.1
13
1446
XM_017018757.1
XP_016874246.1
14
1616
XM_017018758.1
XP_016874247.1
13
1552
XM_017018759.1
XP_016874248.1
12
1377
XM_017018760.1
XP_016874249.1
13
1696
XM_017018761.1
XP_016874250.1
12
1476
XM_017018762.1
XP_016874251.1
12
1476
GenBank mRNA
AF370416.1
-
13
1574
AK055521.1
-
11
1244
AK125113.1
-
12
1486
AK291746.1
-
13
1565
AL832198.1
-
12
1424
AL833272.1
-
13
1606
BC022377.1
-
11
1170
BX647457.1
-
12
1307
CR936626.1
-
13
1614
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
regulation of cell morphogenesis
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-9wt54:80/100.66.77.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0