Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SFTPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000542.3 | NP_000533.3 | 7 | 792 | ||
NM_198843.2 | NP_942140.2 | 7 | 792 | |||
XM_005264487.2 | XP_005264544.1 | 6 | 707 | |||
XM_005264488.3 | XP_005264545.2 | 6 | 672 | |||
XM_005264490.4 | XP_005264547.2 | 6 | 672 | |||
XM_017004702.1 | XP_016860191.1 | 6 | 707 | |||
XM_017004703.1 | XP_016860192.1 | 6 | 672 | |||
GenBank mRNA | AK303817.1 | - | 5 | 580 | ||
AK314947.1 | - | 6 | 672 | |||
AL548140.3 | - | 7 | 791 | |||
BC032785.1 | - | 6 | 659 | 5179408 | ||
DQ317589.1 | - | 6 | 645 | |||
J02761.1 | - | 6 | 669 | |||
M16764.1 | - | 3 | 371 | |||
M19097.1 | - | 2 | 242 |