Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PRKCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316329.1 | NP_001303258.1 | 19 | 2863 | ||
NM_002739.4 | NP_002730.1 | 18 | 2965 | |||
GenBank mRNA | BC047876.1 | - | 18 | 2840 | 5764695 |