Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PSMB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002799.3 | NP_002790.1 | 5 | 512 | ||
GenBank mRNA | AJ420455.1 | - | 2 | 190 | ||
AK098757.1 | - | 5 | 481 | |||
AK223007.1 | - | 5 | 465 | |||
AK223356.1 | - | 5 | 477 | |||
AK314302.1 | - | 5 | 481 | |||
BC000509.2 | - | 5 | 468 | 2822305 | ||
BC008414.1 | - | 5 | 478 | |||
BC017116.2 | - | 4 | 403 | |||
BT007218.1 | - | 5 | 464 | |||
CR456721.1 | - | 5 | 464 | |||
D38048.1 | - | 5 | 478 | |||
GU727627.1 | - | 5 | 481 |