Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002900.2 | NP_002891.1 | 1 | 2302 | ||
GenBank mRNA | AB593121.1 | - | 1 | 2294 | ||
BC039844.1 | - | 1 | 2292 | 5578501 | ||
J03912.1 | - | 1 | 2185 | |||
M22453.1 | - | 1 | 2303 |