Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCL2 SIRNA Assays ›
          Gene Symbol:
          CCL2
          Gene Name:
          C-C motif chemokine ligand 2
          Gene Aliases:
          GDCF-2, HC11, HSMCR30, MCAF, MCP-1, MCP1, SCYA2, SMC-CF
          Chromosome Location:
          Chr.17: 34255277 - 34257203 on Build GRCh38
          UniGene ID:
          Hs.303649
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 142542

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002982.3 NP_002973.1 2 197
          GenBank mRNA AK311960.1 - 2 189
          AV733621.2 - 2 197
          BC009716.1 - 2 180 3901489
          BG530064.1 - 2 210
          BT007329.1 - 2 124
          M24545.1 - 2 193
          M26683.1 - 1 62
          M28226.1 - 2 170
          S69738.1 - 2 155
          S71513.1 - 2 177
          X14768.1 - 2 177

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          angiogenesis
          response to hypoxia
          positive regulation of leukocyte mediated cytotoxicity
          positive regulation of endothelial cell proliferation
          leukocyte migration involved in inflammatory response
          monocyte chemotaxis
          positive regulation of cellular extravasation
          protein phosphorylation
          cellular calcium ion homeostasis
          chemotaxis
          inflammatory response
          humoral immune response
          cytoskeleton organization
          cell adhesion
          signal transduction
          cell surface receptor signaling pathway
          transforming growth factor beta receptor signaling pathway
          G-protein coupled receptor signaling pathway
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          JAK-STAT cascade
          aging
          regulation of cell shape
          response to heat
          response to wounding
          response to mechanical stimulus
          response to bacterium
          organ morphogenesis
          response to gamma radiation
          regulation of vascular endothelial growth factor production
          positive regulation of macrophage chemotaxis
          response to activity
          negative regulation of angiogenesis
          viral genome replication
          cytokine-mediated signaling pathway
          cellular homeostasis
          neutrophil chemotaxis
          organ regeneration
          lipopolysaccharide-mediated signaling pathway
          response to progesterone
          positive regulation of tumor necrosis factor production
          cellular response to insulin stimulus
          positive regulation of collagen biosynthetic process
          response to vitamin B3
          negative regulation of glial cell apoptotic process
          helper T cell extravasation
          cellular response to drug
          cellular response to macrophage colony-stimulating factor stimulus
          cellular response to platelet-derived growth factor stimulus
          PERK-mediated unfolded protein response
          response to amino acid
          protein kinase B signaling
          negative regulation of neuron apoptotic process
          positive regulation of GTPase activity
          astrocyte cell migration
          cellular response to fibroblast growth factor stimulus
          response to ethanol
          response to antibiotic
          vascular endothelial growth factor receptor signaling pathway
          macrophage chemotaxis
          lymphocyte chemotaxis
          positive regulation of inflammatory response
          positive regulation of synaptic transmission
          positive regulation of T cell activation
          positive regulation of nitric-oxide synthase biosynthetic process
          maternal process involved in female pregnancy
          maternal process involved in parturition
          chemokine-mediated signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          cellular response to retinoic acid
          cellular response to ATP
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to interleukin-6
          cellular response to tumor necrosis factor
          cellular response to high density lipoprotein particle stimulus
          cellular response to organic cyclic compound
          cellular response to dexamethasone stimulus
          positive regulation of monocyte chemotaxis
          positive regulation of immune complex clearance by monocytes and macrophages
          positive regulation of calcium ion import
          positive regulation of protein targeting to membrane
          positive regulation of apoptotic cell clearance
          negative regulation of natural killer cell chemotaxis
          protein kinase activity
          receptor binding
          chemokine activity
          heparin binding
          CCR2 chemokine receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0