Search
Search

SMARCD3| Gene Symbol: SMARCD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003801.1 | NP_001003801.1 | 13 | 1557 | ||
| NM_001003802.1 | NP_001003802.1 | 14 | 1546 | |||
| NM_003078.3 | NP_003069.2 | 14 | 1585 | |||
| XM_011516521.2 | XP_011514823.1 | 13 | 1664 | |||
| XM_017012556.1 | XP_016868045.1 | 13 | 1400 | |||
| GenBank mRNA | AK127595.1 | - | 14 | 2093 | ||
| AY450430.1 | - | 13 | 1861 | |||
| AY450431.1 | - | 14 | 1546 | |||
| BC002628.2 | - | 14 | 1546 | 3608512 | ||
| BC110350.1 | - | 13 | 1476 | 5212363 | ||
| BX648385.1 | - | 12 | 2308 | |||