Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNAP25 SIRNA Assays ›
          Gene Symbol:
          SNAP25
          Gene Name:
          synaptosome associated protein 25
          Gene Aliases:
          CMS18, RIC-4, RIC4, SEC9, SNAP, SNAP-25, SUP, bA416N4.2, dJ1068F16.2
          Chromosome Location:
          Chr.20: 10218694 - 10307420 on Build GRCh38
          UniGene ID:
          Hs.167317
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 142673

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNAP25
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001322902.1 NP_001309831.1 8 1047
          NM_001322903.1 NP_001309832.1 9 1143
          NM_001322904.1 NP_001309833.1 9 1113
          NM_001322905.1 NP_001309834.1 9 1076
          NM_001322906.1 NP_001309835.1 9 1070
          NM_001322907.1 NP_001309836.1 10 1271
          NM_001322908.1 NP_001309837.1 10 1194
          NM_001322909.1 NP_001309838.1 9 1061
          NM_001322910.1 NP_001309839.1 9 1168
          NM_003081.4 NP_003072.2 8 1016
          NM_130811.3 NP_570824.1 8 1016
          XM_005260808.4 XP_005260865.1 8 1094
          XM_017028021.1 XP_016883510.1 9 946
          XM_017028022.1 XP_016883511.1 9 1038
          GenBank mRNA AK026732.1 - 3 1187
          AK090857.1 - 3 1984
          AK094560.1 - 9 1102
          AK098347.1 - 9 744
          AK098767.1 - 8 886
          AK223617.1 - 8 978
          AK289647.1 - 10 1177
          BC010647.1 - 8 905 3867544
          BX647443.1 - 8 888
          D21267.1 - 8 978
          L19760.1 - 8 861
          L19761.1 - 8 861

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter uptake
          chemical synaptic transmission
          neurotransmitter secretion
          locomotory behavior
          associative learning
          regulation of neuron projection development
          glutamate secretion
          synaptic vesicle exocytosis
          synaptic vesicle docking
          synaptic vesicle priming
          synaptic vesicle fusion to presynaptic active zone membrane
          regulation of insulin secretion
          long-term synaptic potentiation
          SNAP receptor activity
          protein binding
          syntaxin-1 binding
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mvhs5:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0