Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAPK10 SIRNA Assays ›
          Gene Symbol:
          MAPK10
          Gene Name:
          mitogen-activated protein kinase 10
          Gene Aliases:
          JNK3, JNK3A, PRKM10, SAPK1b, p493F12, p54bSAPK
          Chromosome Location:
          Chr.4: 86012296 - 86594117 on Build GRCh38
          UniGene ID:
          Hs.125503
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 143164

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MAPK10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001318067.1 NP_001304996.1 4 416
          NM_001318068.1 NP_001304997.1 5 570
          NM_001318069.1 NP_001304998.1 5 981
          NM_002753.4 NP_002744.1 5 517
          NM_138980.3 NP_620446.1 5 1008
          NM_138982.3 NP_620448.1 5 981
          XM_005263129.2 XP_005263186.1 5 1078
          XM_005263130.2 XP_005263187.1 5 474
          XM_005263131.2 XP_005263188.1 5 875
          XM_005263135.3 XP_005263192.1 5 875
          XM_006714268.2 XP_006714331.1 5 748
          XM_006714269.2 XP_006714332.1 5 875
          XM_011532117.2 XP_011530419.1 5 581
          XM_011532118.2 XP_011530420.1 5 674
          XM_011532120.2 XP_011530422.1 5 644
          XM_011532121.2 XP_011530423.1 5 510
          XM_017008419.1 XP_016863908.1 6 771
          XM_017008420.1 XP_016863909.1 5 1205
          XM_017008421.1 XP_016863910.1 5 801
          XM_017008422.1 XP_016863911.1 3 301
          XM_017008423.1 XP_016863912.1 5 493
          XM_017008424.1 XP_016863913.1 5 828
          XM_017008425.1 XP_016863914.1 5 501
          XM_017008426.1 XP_016863915.1 5 829
          XM_017008427.1 XP_016863916.1 5 902
          XM_017008428.1 XP_016863917.1 5 644
          XM_017008429.1 XP_016863918.1 5 1205
          XM_017008430.1 XP_016863919.1 5 583
          XM_017008431.1 XP_016863920.1 5 801
          XM_017008432.1 XP_016863921.1 5 474
          XM_017008433.1 XP_016863922.1 5 902
          XM_017008434.1 XP_016863923.1 5 858
          XM_017008435.1 XP_016863924.1 5 748
          XM_017008436.1 XP_016863925.1 5 1232
          XM_017008437.1 XP_016863926.1 5 510
          XM_017008438.1 XP_016863927.1 5 501
          XM_017008439.1 XP_016863928.1 5 829
          XM_017008440.1 XP_016863929.1 5 902
          XM_017008441.1 XP_016863930.1 5 902
          XM_017008442.1 XP_016863931.1 7 1223
          XM_017008443.1 XP_016863932.1 7 874
          XM_017008444.1 XP_016863933.1 7 1196
          XM_017008445.1 XP_016863934.1 4 705
          XM_017008446.1 XP_016863935.1 2 1397
          XM_017008447.1 XP_016863936.1 5 902
          XM_017008448.1 XP_016863937.1 5 875
          XM_017008449.1 XP_016863938.1 5 902
          XM_017008450.1 XP_016863939.1 5 875
          XM_017008451.1 XP_016863940.1 4 642
          XM_017008452.1 XP_016863941.1 5 902
          XM_017008453.1 XP_016863942.1 3 301
          XM_017008454.1 XP_016863943.1 5 581
          GenBank mRNA AK057723.1 - 1 362
          AK124791.1 - 5 1233
          AK225974.1 - 5 562
          AK294125.1 - 1 32
          AK307129.1 - 6 694
          BC022492.1 - 5 814
          BC035057.1 - 5 826 5261221
          BC051731.1 - 6 686 4838495
          BC065516.1 - 1 37 5721954
          U07620.1 - 5 518
          U34819.1 - 4 362
          U34820.1 - 4 386

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          protein phosphorylation
          signal transduction
          JNK cascade
          JUN phosphorylation
          response to light stimulus
          Fc-epsilon receptor signaling pathway
          regulation of circadian rhythm
          rhythmic process
          regulation of sequence-specific DNA binding transcription factor activity
          JUN kinase activity
          MAP kinase kinase activity
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0