Search Thermo Fisher Scientific
Gene Symbol: PEX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000287.3 | NP_000278.3 | 17 | 3014 | ||
NM_001316313.1 | NP_001303242.1 | 17 | 2750 | |||
NR_133009.1 | 15 | 2767 | ||||
XM_011514661.1 | XP_011512963.1 | 16 | 2930 | |||
GenBank mRNA | AB051076.1 | - | 17 | 2921 | ||
AB051077.1 | - | 17 | 2657 | |||
AB051078.1 | - | 15 | 2674 | |||
AB208906.1 | - | 13 | 3608 | |||
AK314237.1 | - | 17 | 2950 | |||
BC033487.1 | - | 17 | 2929 | |||
BC048331.1 | - | 17 | 2937 | 5189685 | ||
D83703.1 | - | 17 | 2991 |