Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT2 SIRNA Assays ›
Gene Symbol:
SEPT2
Gene Name:
septin 2
Gene Aliases:
DIFF6, NEDD-5, NEDD5, Pnutl3, hNedd5
Chromosome Location:
Chr.2: 241315187 - 241354027 on Build GRCh38
UniGene ID:
Hs.721234
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
143876
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008491.2
NP_001008491.1
9
1228
NM_001008492.2
NP_001008492.1
9
910
NM_001282972.1
NP_001269901.1
9
959
NM_001282973.1
NP_001269902.1
12
1183
NM_001321029.1
NP_001307958.1
9
944
NM_001321030.1
NP_001307959.1
9
942
NM_001321031.1
NP_001307960.1
10
1045
NM_001321032.1
NP_001307961.1
8
825
NM_001321033.1
NP_001307962.1
10
1061
NM_001321034.1
NP_001307963.1
10
1060
NM_001321035.1
NP_001307964.1
9
926
NM_004404.4
NP_004395.1
8
825
NM_006155.2
NP_006146.1
8
1127
XM_011511237.1
XP_011509539.1
8
791
XM_011511238.1
XP_011509540.1
7
659
XM_011511240.1
XP_011509542.1
7
668
XM_017004189.1
XP_016859678.1
10
1020
XM_017004190.1
XP_016859679.1
10
955
XM_017004191.1
XP_016859680.1
10
1020
XM_017004192.1
XP_016859681.1
10
1020
XM_017004193.1
XP_016859682.1
8
825
XM_017004194.1
XP_016859683.1
10
975
XM_017004195.1
XP_016859684.1
10
1179
XM_017004196.1
XP_016859685.1
10
1061
XM_017004197.1
XP_016859686.1
9
876
XM_017004198.1
XP_016859687.1
9
938
XM_017004199.1
XP_016859688.1
10
977
XM_017004200.1
XP_016859689.1
9
940
XM_017004201.1
XP_016859690.1
9
1062
XM_017004202.1
XP_016859691.1
8
1044
XM_017004203.1
XP_016859692.1
9
868
XM_017004204.1
XP_016859693.1
8
783
XM_017004205.1
XP_016859694.1
10
975
XM_017004206.1
XP_016859695.1
9
876
XM_017004207.1
XP_016859696.1
6
1274
XM_017004208.1
XP_016859697.1
9
1484
XM_017004209.1
XP_016859698.1
8
1371
XM_017004210.1
XP_016859699.1
8
1415
XM_017004211.1
XP_016859700.1
7
1268
XM_017004212.1
XP_016859701.1
7
1520
GenBank mRNA
AF038404.1
-
8
764
AK294563.1
-
9
867
BC014455.1
-
9
1107
4906575
BC033559.1
-
12
1161
4821654
BX648000.1
-
9
844
D28540.1
-
7
748
D63878.1
-
8
944
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of L-glutamate transport
mitotic nuclear division
smoothened signaling pathway
neuron projection development
regulation of protein localization
cilium assembly
regulation of catalytic activity
cell division
cell-cell adhesion
protein binding
GTP binding
enzyme regulator activity
protein complex scaffold
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-sftpw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0