Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATXN3 SIRNA Assays ›
Gene Symbol:
ATXN3
ATXN8
Gene Name:
ataxin 3
ataxin 8
Gene Aliases:
AT3, ATX3, JOS, MJD, MJD1, SCA3
ATXN3
Chromosome Location:
Chr.14: 92058552 - 92106621 on Build GRCh38
Chromosome Location:
-
UniGene ID:
Hs.532632
Hs.532632
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
143939
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127696.1
NP_001121168.1
7
690
NM_001127697.2
NP_001121169.2
6
582
NM_001164779.1
NP_001158251.1
4
372
NM_001164780.1
NP_001158252.1
5
439
NM_001164781.1
NP_001158253.1
6
525
NM_004993.5
NP_004984.2
8
735
NM_030660.4
NP_109376.1
7
570
NR_028453.1
7
649
NR_028454.1
6
484
NR_028455.1
7
649
NR_028456.1
6
484
NR_028457.1
8
740
NR_028458.1
7
626
NR_028459.1
8
735
NR_028461.1
7
647
NR_028462.1
7
626
NR_028463.1
3
284
NR_028464.1
6
582
NR_028465.1
6
604
NR_028466.1
3
284
NR_028467.1
7
650
NR_028469.1
6
484
NR_031765.1
2
151
GenBank mRNA
AB050194.1
-
7
559
AB209309.1
-
11
AK225884.1
-
8
674
AK307720.1
-
5
400
AK314919.1
-
8
687
BC022245.1
-
7
618
BC033711.1
-
8
678
4393766
BC095402.1
-
8
695
30347215
BU190081.1
-
7
669
BU663113.1
-
5
489
GQ176434.1
-
8
687
GQ176435.1
-
8
688
GQ176436.1
-
6
535
GQ176437.1
-
6
535
GQ176438.1
-
7
602
GQ176440.1
-
7
523
GQ176441.1
-
7
602
GQ176442.1
-
6
437
GQ176443.1
-
7
602
GQ176445.1
-
6
437
GQ176446.1
-
6
434
GQ176448.1
-
8
688
GQ176451.1
-
6
437
GQ176452.1
-
7
688
GQ176453.1
-
7
578
GQ176454.1
-
8
688
GQ176455.1
-
7
523
GQ176456.1
-
6
437
GQ176457.1
-
6
535
GQ176458.1
-
7
602
GQ176459.1
-
4
325
GQ176462.1
-
8
688
GQ176463.1
-
7
602
GQ176464.1
-
7
602
GQ176465.1
-
3
237
GQ176466.1
-
6
437
GQ176467.1
-
7
523
GQ176469.1
-
6
535
GQ176470.1
-
6
557
GQ176474.1
-
6
535
GQ176475.1
-
6
437
GQ176476.1
-
8
688
GQ176477.1
-
7
602
GQ176481.1
-
7
602
GQ176483.1
-
7
602
GQ176484.1
-
7
602
GQ176485.1
-
6
534
GQ176486.1
-
7
643
GQ176488.1
-
8
688
GQ176489.1
-
7
643
GQ176490.1
-
6
569
GQ176492.1
-
7
602
GQ176493.1
-
6
534
GQ176495.1
-
6
535
GQ176496.1
-
6
437
GQ176497.1
-
7
602
GQ176498.1
-
7
523
GQ176499.1
-
8
688
GQ176500.1
-
8
689
GQ176501.1
-
7
579
GQ176502.1
-
7
602
GQ176503.1
-
6
436
GQ176504.1
-
8
688
GQ176506.1
-
6
437
GQ176507.1
-
7
601
GQ176508.1
-
8
688
GQ176511.1
-
6
535
GQ176512.1
-
8
685
GQ176513.1
-
7
602
GQ176514.1
-
5
392
GQ176515.1
-
6
435
GQ176516.1
-
8
688
GQ176517.1
-
3
237
GQ176518.1
-
3
237
GQ176519.1
-
7
601
GQ176520.1
-
5
392
GQ176521.1
-
6
437
GQ176522.1
-
6
437
GQ176523.1
-
8
688
GQ176524.1
-
7
602
GQ176525.1
-
6
437
GQ176526.1
-
6
478
GQ176527.1
-
7
602
GQ176528.1
-
7
602
GQ176529.1
-
7
599
GQ176530.1
-
7
602
GQ176531.1
-
8
688
GQ176532.1
-
7
688
GQ176533.1
-
8
688
GQ176534.1
-
7
599
GQ176535.1
-
437
GQ176536.1
-
7
523
GQ176537.1
-
7
602
GQ176538.1
-
8
688
GQ176539.1
-
7
523
GQ176540.1
-
7
576
GQ176542.1
-
4
324
GQ176543.1
-
6
437
GQ176545.1
-
4
325
GQ176546.1
-
7
602
GQ176548.1
-
8
688
GQ176549.1
-
6
437
GQ176550.1
-
8
688
GQ176551.1
-
8
686
GQ176552.1
-
8
687
GQ176556.1
-
6
437
GQ176559.1
-
6
437
GQ176560.1
-
8
688
GQ176563.1
-
8
688
GQ176565.1
-
8
688
GQ176566.1
-
7
523
GQ176567.1
-
8
688
GQ176568.1
-
8
688
GQ176569.1
-
7
599
GQ176570.1
-
8
688
GQ176571.1
-
6
437
GQ176572.1
-
7
602
GQ176573.1
-
7
523
GQ176576.1
-
7
600
GQ176577.1
-
8
688
GQ176578.1
-
8
688
GQ176579.1
-
7
579
GQ176580.1
-
8
688
GQ176583.1
-
2
104
GQ176585.1
-
6
436
GQ176586.1
-
7
602
GQ176587.1
-
6
437
GQ176588.1
-
7
602
GQ176589.1
-
8
688
GQ176590.1
-
2
104
GQ176591.1
-
6
437
GQ176592.1
-
7
522
GQ176593.1
-
9
774
GQ176594.1
-
8
688
GQ176596.1
-
7
602
GQ176597.1
-
8
688
GQ176598.1
-
6
437
GQ176599.1
-
8
688
GQ176600.1
-
7
602
GQ176601.1
-
6
437
GQ176602.1
-
8
706
GQ176603.1
-
6
437
GQ176604.1
-
6
437
GQ176605.1
-
7
602
GQ176607.1
-
7
602
GQ176608.1
-
6
535
GQ176610.1
-
7
602
GQ176612.1
-
8
688
GQ176614.1
-
5
392
GQ176615.1
-
688
GQ176616.1
-
8
688
GQ176617.1
-
7
602
GQ176618.1
-
514
GQ176619.1
-
8
688
GQ176620.1
-
7
602
GQ176621.1
-
8
688
GQ176622.1
-
8
688
GQ176623.1
-
6
437
GQ176624.1
-
7
621
GQ176625.1
-
325
GQ176626.1
-
6
437
GQ176627.1
-
4
325
GQ176628.1
-
688
GQ176629.1
-
602
GQ176630.1
-
7
602
GQ176631.1
-
6
535
GQ176632.1
-
8
688
GQ176633.1
-
4
325
GQ176635.1
-
437
GQ176636.1
-
5
509
GQ176637.1
-
6
437
GQ176638.1
-
6
437
GQ176639.1
-
6
534
GQ176640.1
-
8
688
GQ176641.1
-
408
GQ176642.1
-
8
688
GQ176643.1
-
437
GQ176645.1
-
8
688
GQ176646.1
-
8
688
GQ176647.1
-
437
GQ176648.1
-
7
602
GQ176649.1
-
7
602
GQ176650.1
-
3
237
GQ176653.1
-
5
392
GQ176654.1
-
7
602
GQ176655.1
-
6
532
GQ176656.1
-
6
530
GQ176657.1
-
6
437
GQ176658.1
-
8
688
GQ176659.1
-
602
GQ176660.1
-
7
602
GQ176661.1
-
5
392
GQ176662.1
-
7
602
GQ176664.1
-
7
602
GQ176665.1
-
7
523
GQ176666.1
-
5
392
GQ176668.1
-
8
685
GQ176669.1
-
602
GQ176670.1
-
5
490
GQ176671.1
-
688
GQ176673.1
-
8
688
GQ176674.1
-
8
688
GQ176675.1
-
7
523
GQ176676.1
-
6
437
GQ176677.1
-
8
688
GQ176678.1
-
7
602
GQ176679.1
-
7
523
GQ176681.1
-
5
447
GQ176682.1
-
7
602
GQ176684.1
-
8
688
GQ176685.1
-
8
687
GQ176686.1
-
688
GQ176687.1
-
8
688
GQ176688.1
-
6
534
GQ176689.1
-
5
392
GQ176690.1
-
7
602
GQ176691.1
-
6
437
GQ176692.1
-
8
688
GQ176693.1
-
688
GQ176695.1
-
6
535
GQ176696.1
-
7
602
GQ176697.1
-
8
688
GQ176699.1
-
688
GQ176700.1
-
7
523
GQ176702.1
-
5
392
GQ176703.1
-
8
688
GQ176704.1
-
8
688
GQ176705.1
-
4
325
GQ176706.1
-
7
602
GQ176707.1
-
434
GQ176708.1
-
9
774
GQ176709.1
-
599
GQ176710.1
-
4
325
GQ176711.1
-
5
392
GQ176712.1
-
7
643
GQ176713.1
-
437
GQ176714.1
-
8
688
GQ176715.1
-
8
610
GQ176716.1
-
6
437
GQ176717.1
-
602
GQ176719.1
-
688
GQ176720.1
-
6
437
GQ176721.1
-
8
688
GQ176722.1
-
7
602
GQ176723.1
-
5
392
GQ176724.1
-
6
535
GQ176725.1
-
8
688
GQ176726.1
-
6
437
GQ176727.1
-
4
325
GQ176728.1
-
8
688
GQ176729.1
-
6
437
GQ176732.1
-
437
GQ176733.1
-
7
602
GQ176734.1
-
535
GQ176735.1
-
8
688
GQ176736.1
-
437
GQ176737.1
-
8
688
GQ176738.1
-
7
602
GQ176740.1
-
6
437
GQ176742.1
-
370
GQ176743.1
-
6
437
GQ176744.1
-
7
643
GQ176745.1
-
688
GQ176746.1
-
8
688
GQ176747.1
-
688
GQ176748.1
-
6
437
GQ176749.1
-
8
688
GQ176750.1
-
7
523
GQ176751.1
-
437
GQ176753.1
-
5
392
GQ176754.1
-
7
523
GQ176755.1
-
7
602
GQ176756.1
-
8
688
GQ176757.1
-
6
535
GQ176758.1
-
6
437
GQ176759.1
-
8
688
GQ176760.1
-
7
602
GQ176761.1
-
5
370
GQ176762.1
-
688
GQ176763.1
-
8
688
GQ176764.1
-
437
GQ176765.1
-
6
437
GQ176766.1
-
8
688
GQ176767.1
-
7
602
GQ176768.1
-
6
437
GQ176769.1
-
5
490
GQ176771.1
-
8
688
GQ176772.1
-
6
535
GQ176774.1
-
6
437
GQ176776.1
-
688
GQ176778.1
-
5
392
GQ176779.1
-
6
535
GQ176780.1
-
6
437
GQ176781.1
-
6
535
GQ176782.1
-
6
535
GQ176784.1
-
6
535
GQ176785.1
-
5
370
GQ176786.1
-
6
535
GQ176787.1
-
8
688
GQ176788.1
-
6
437
GQ176789.1
-
6
535
GQ176790.1
-
6
534
GQ176791.1
-
8
688
GQ176792.1
-
6
532
GQ176793.1
-
6
535
GQ176794.1
-
392
GQ176795.1
-
6
437
GQ176796.1
-
602
GQ176797.1
-
7
602
GQ176798.1
-
602
GQ176799.1
-
370
GQ176800.1
-
688
GQ176801.1
-
601
GQ176802.1
-
535
GQ176803.1
-
602
GQ176804.1
-
325
GQ176805.1
-
8
664
GQ176806.1
-
8
687
GQ176807.1
-
7
602
GQ176808.1
-
688
GQ176809.1
-
5
490
GQ176810.1
-
7
600
GQ176811.1
-
8
689
GQ176812.1
-
6
535
GQ176813.1
-
7
523
GQ176814.1
-
6
434
GQ176815.1
-
6
536
GQ176816.1
-
602
GQ176817.1
-
4
324
GQ176818.1
-
7
643
GQ176819.1
-
7
602
GQ176820.1
-
8
688
GQ176821.1
-
6
436
GQ176822.1
-
7
523
GQ176823.1
-
7
579
GQ176824.1
-
8
688
GQ176826.1
-
8
665
GQ176827.1
-
8
688
GQ176828.1
-
6
437
GQ176829.1
-
8
688
GQ176830.1
-
659
GQ176831.1
-
688
GQ176832.1
-
6
437
GQ176834.1
-
601
GQ176835.1
-
7
522
GQ176836.1
-
7
523
GQ176837.1
-
8
688
GQ176838.1
-
8
688
GQ176839.1
-
437
GQ176840.1
-
6
437
GQ176841.1
-
8
688
GQ176842.1
-
602
GQ176843.1
-
6
436
GQ176844.1
-
6
450
GQ176845.1
-
7
601
GQ176846.1
-
8
688
S75313.1
-
8
701
U64820.1
-
8
723
U64821.1
-
8
686
U64822.1
-
7
663
Gene Symbol: ATXN8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
GQ176777.1
-
370
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
nucleotide-excision repair
transcription, DNA-templated
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
misfolded or incompletely synthesized protein catabolic process
chemical synaptic transmission
nervous system development
regulation of cell-substrate adhesion
protein deubiquitination
actin cytoskeleton organization
cellular response to heat
monoubiquitinated protein deubiquitination
exploration behavior
proteasome-mediated ubiquitin-dependent protein catabolic process
intermediate filament cytoskeleton organization
protein K63-linked deubiquitination
histone H3 deacetylation
protein K48-linked deubiquitination
cellular response to misfolded protein
positive regulation of ERAD pathway
protein localization to cytosolic proteasome complex involved in ERAD pathway
RNA polymerase II regulatory region DNA binding
histone deacetylase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
omega peptidase activity
ubiquitin protein ligase binding
identical protein binding
ATPase binding
Lys63-specific deubiquitinase activity
Lys48-specific deubiquitinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0