Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NUCB2 SIRNA Assays ›
          Gene Symbol:
          NUCB2
          Gene Name:
          nucleobindin 2
          Gene Aliases:
          HEL-S-109, NEFA
          Chromosome Location:
          Chr.11: 17260340 - 17331881 on Build GRCh38
          UniGene ID:
          Hs.654599
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 143960

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NUCB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005013.2 NP_005004.1 9 1055
          XM_017017804.1 XP_016873293.1 11 1290
          XM_017017805.1 XP_016873294.1 10 1206
          XM_017017806.1 XP_016873295.1 10 1149
          XM_017017807.1 XP_016873296.1 10 1160
          XM_017017808.1 XP_016873297.1 9 1094
          XM_017017809.1 XP_016873298.1 9 1091
          XM_017017810.1 XP_016873299.1 11 1561
          XM_017017811.1 XP_016873300.1 11 1470
          XM_017017812.1 XP_016873301.1 9 1055
          XM_017017813.1 XP_016873302.1 10 1154
          XM_017017814.1 XP_016873303.1 10 1147
          XM_017017815.1 XP_016873304.1 12 1554
          XM_017017816.1 XP_016873305.1 10 1324
          XM_017017817.1 XP_016873306.1 10 1249
          XM_017017818.1 XP_016873307.1 8 900
          XM_017017819.1 XP_016873308.1 8 845
          XM_017017820.1 XP_016873309.1 11 1290
          XM_017017821.1 XP_016873310.1 11 1286
          XM_017017822.1 XP_016873311.1 9 1058
          XM_017017823.1 XP_016873312.1 10 1162
          XM_017017824.1 XP_016873313.1 10 1160
          XM_017017825.1 XP_016873314.1 9 1078
          XM_017017826.1 XP_016873315.1 9 1091
          XM_017017827.1 XP_016873316.1 11 1517
          XM_017017828.1 XP_016873317.1 10 1147
          XM_017017829.1 XP_016873318.1 8 923
          GenBank mRNA AB231311.1 - 7 810
          AF052642.1 - 7 810
          AF052643.1 - 7 810
          AF052644.1 - 7 810
          AF450266.1 - 9 1024
          AK097398.1 - 11 1479
          AK128739.1 - 13 2722
          AK291507.1 - 9 1085
          FJ224327.1 - 9 1029
          X76732.1 - 9 1029

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Process(es)

          DNA binding
          calcium ion binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline