Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IFNB1 SIRNA Assays ›
          Gene Symbol:
          IFNB1
          Gene Name:
          interferon beta 1
          Gene Aliases:
          IFB, IFF, IFN-beta, IFNB
          Chromosome Location:
          Chr.9: 21077105 - 21077963 on Build GRCh38
          UniGene ID:
          Hs.93177
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 144950

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IFNB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002176.3 NP_002167.1 1 423
          GenBank mRNA AB451323.1 - 1 329
          AB451452.1 - 1 329
          AB451491.1 - 1 266
          BC069314.1 - 1 329 7262165
          BC096150.3 - 1 329 40002940
          BC096151.2 - 1 329 40002941
          BC096152.2 - 1 329 40002943
          BC096153.2 - 1 329 40002945
          M25460.1 - 1 403
          M28622.1 - 1 404
          V00546.1 - 1 393
          V00547.1 - 1 329

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          B cell activation involved in immune response
          natural killer cell activation involved in immune response
          humoral immune response
          cell surface receptor signaling pathway
          blood coagulation
          response to virus
          natural killer cell activation
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          cellular response to interferon-beta
          B cell proliferation
          defense response to bacterium
          response to exogenous dsRNA
          negative regulation of viral genome replication
          positive regulation of innate immune response
          regulation of MHC class I biosynthetic process
          negative regulation of T cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          defense response to virus
          type I interferon signaling pathway
          regulation of type I interferon-mediated signaling pathway
          cellular response to exogenous dsRNA
          cellular response to dexamethasone stimulus
          cellular response to virus
          negative regulation of T-helper 2 cell cytokine production
          positive regulation of apoptotic signaling pathway
          cytokine activity
          type I interferon receptor binding
          chloramphenicol O-acetyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0