Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ISL1 SIRNA Assays ›
          Gene Symbol:
          ISL1
          Gene Name:
          ISL LIM homeobox 1
          Gene Aliases:
          ISLET1, Isl-1
          Chromosome Location:
          Chr.5: 51383124 - 51394730 on Build GRCh38
          UniGene ID:
          Hs.505
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 144976

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ISL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002202.2 NP_002193.2 4, 5 1306
          XM_011543380.2 XP_011541682.1 3, 4 2424
          GenBank mRNA AK290742.1 - 4, 5 1024
          BC031213.1 - 4, 5 1307 5269369
          S70721.1 - 4, 5 763
          U07559.1 - 4, 5 997

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          neural crest cell migration
          secondary heart field specification
          outflow tract septum morphogenesis
          outflow tract morphogenesis
          endocardial cushion morphogenesis
          cardiac right ventricle morphogenesis
          regulation of secondary heart field cardioblast proliferation
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          positive regulation of cell proliferation
          positive regulation of vascular endothelial growth factor production
          spinal cord motor neuron cell fate specification
          spinal cord motor neuron differentiation
          visceral motor neuron differentiation
          trigeminal nerve development
          pituitary gland development
          pancreas development
          axon regeneration
          retinal ganglion cell axon guidance
          positive regulation of insulin secretion
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-1 alpha production
          positive regulation of interleukin-1 beta production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          negative regulation of intracellular estrogen receptor signaling pathway
          positive regulation of histone acetylation
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of DNA binding
          negative regulation of neuron apoptotic process
          negative regulation of neuron differentiation
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          neuron fate specification
          mesenchymal cell differentiation
          sensory system development
          peripheral nervous system neuron axonogenesis
          negative regulation of inflammatory response
          ventricular cardiac muscle tissue morphogenesis
          pharyngeal system development
          cardiac muscle cell myoblast differentiation
          innervation
          atrial septum morphogenesis
          cardiac cell fate determination
          cellular response to glucocorticoid stimulus
          positive regulation of granulocyte colony-stimulating factor production
          negative regulation of protein homodimerization activity
          negative regulation of canonical Wnt signaling pathway
          positive regulation of macrophage colony-stimulating factor production
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II activating transcription factor binding
          RNA polymerase II transcription coactivator activity
          enhancer sequence-specific DNA binding
          chromatin binding
          protein binding
          zinc ion binding
          ligand-dependent nuclear receptor binding
          estrogen receptor binding
          bHLH transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0