Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GGT5 SIRNA Assays ›
          Gene Symbol:
          GGT5
          Gene Name:
          gamma-glutamyltransferase 5
          Gene Aliases:
          GGL, GGT 5, GGT-REL, GGTLA1
          Chromosome Location:
          Chr.22: 24219654 - 24245142 on Build GRCh38
          UniGene ID:
          Hs.437156
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 145067

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GGT5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099781.2 NP_001093251.1 8, 9 1642
          NM_001099782.2 NP_001093252.1 7, 8 1546
          NM_001302464.1 NP_001289393.1 7, 8 1546
          NM_001302465.1 NP_001289394.1 6, 7 1411
          NM_004121.3 NP_004112.2 8, 9 1642
          XM_005261557.2 XP_005261614.1 8, 9 1578
          XM_005261558.2 XP_005261615.1 8, 9 1578
          XM_011530133.1 XP_011528435.1 8, 9 1578
          XM_011530134.1 XP_011528436.1 8, 9 1578
          XM_011530135.1 XP_011528437.1 7, 8 1482
          XM_011530136.1 XP_011528438.1 8, 9 1578
          XM_011530137.2 XP_011528439.1 8, 9 1578
          XM_017028768.1 XP_016884257.1 7, 8 1482
          XM_017028769.1 XP_016884258.1 7, 8 1482
          GenBank mRNA AK225661.1 - 7, 8 1482
          AK226019.1 - 7, 8 1482
          AK292006.1 - 8, 9 1424
          AK293683.1 - 6, 7 1351
          AK297869.1 - 9, 10 1417
          AK312401.1 - 8, 9 1578
          BC011362.1 - 8, 9 1631 4179497
          BC073999.1 - 8, 9 1559 6198113
          BT009808.1 - 8, 9 1225
          CT841518.1 - 8, 9 1383
          M64099.1 - 8, 9 1566

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_019235, U76252, BC087018.
          This siRNA also targets the following transcripts in Mouse: NM_011820, AF077765, AK029138, BC113775, AK170326, and others.

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          cellular amino acid metabolic process
          leukotriene metabolic process
          glutathione metabolic process
          glutathione biosynthetic process
          inflammatory response
          leukotriene biosynthetic process
          gamma-glutamyltransferase activity
          glutathione hydrolase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0