Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAMK2B SIRNA Assays ›
Gene Symbol:
CAMK2B
Gene Name:
calcium/calmodulin dependent protein kinase II beta
Gene Aliases:
CAM2, CAMK2, CAMKB
Chromosome Location:
Chr.7: 44217150 - 44325994 on Build GRCh38
UniGene ID:
Hs.351887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
145667
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAMK2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220.4
NP_001211.3
24
2354
NM_172078.2
NP_742075.1
21
1982
NM_172079.2
NP_742076.1
20
1910
NM_172080.2
NP_742077.1
20
1907
NM_172081.2
NP_742078.1
19
1865
NM_172082.2
NP_742079.1
20
1832
NM_172083.2
NP_742080.1
18
1793
NM_172084.2
NP_742081.1
16
1703
XM_005249862.3
XP_005249919.1
20
1813
XM_005249864.3
XP_005249921.1
19
1714
XM_006715781.2
XP_006715844.1
22
2110
XM_006715784.2
XP_006715847.1
20
1786
XM_011515547.1
XP_011513849.1
25
2359
XM_011515549.1
XP_011513851.1
24
2314
XM_011515550.1
XP_011513852.1
24
2287
XM_011515551.1
XP_011513853.1
24
2287
XM_011515552.1
XP_011513854.1
24
2284
XM_011515553.1
XP_011513855.1
24
2245
XM_011515554.1
XP_011513856.1
23
2239
XM_011515555.1
XP_011513857.1
24
2230
XM_011515556.1
XP_011513858.1
23
2215
XM_011515557.1
XP_011513859.1
23
2212
XM_011515559.1
XP_011513861.1
25
2268
XM_017012660.1
XP_016868149.1
23
2242
XM_017012661.1
XP_016868150.1
23
2200
XM_017012662.1
XP_016868151.1
22
2041
XM_017012663.1
XP_016868152.1
24
2196
XM_017012664.1
XP_016868153.1
19
1738
XM_017012665.1
XP_016868154.1
19
1711
XM_017012666.1
XP_016868155.1
21
1767
XM_017012667.1
XP_016868156.1
20
1722
XM_017012668.1
XP_016868157.1
20
1695
XM_017012669.1
XP_016868158.1
20
1695
XM_017012670.1
XP_016868159.1
19
1650
GenBank mRNA
AJ252236.1
-
19
1742
AJ252237.1
-
18
1593
AK222703.1
-
21
1806
BC019070.2
-
19
1859
5014712
DB566810.1
-
3
446
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein phosphorylation
signal transduction
positive regulation of neuron projection development
regulation of skeletal muscle adaptation
positive regulation of phospholipase A2 activity
positive regulation of GTPase activity
protein autophosphorylation
regulation of long-term neuronal synaptic plasticity
cell projection morphogenesis
regulation of synapse structural plasticity
regulation of calcium ion transport
interferon-gamma-mediated signaling pathway
regulation of dendritic spine development
positive regulation of dendritic spine morphogenesis
positive regulation of synapse maturation
regulation of cellular response to heat
actin binding
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
Ras guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
protein kinase binding
protein homodimerization activity
phospholipase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ntzfc:80/100.66.76.85:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0