Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001299.5 | NP_001290.2 | 3, 4 | 455 | ||
NM_001308341.1 | NP_001295270.1 | 4, 5 | 730 | |||
NM_001308342.1 | NP_001295271.1 | 4, 5 | 721 | |||
XM_005259741.2 | XP_005259798.1 | 3, 4 | 304 | |||
XM_017026289.1 | XP_016881778.1 | 3, 4 | 627 | |||
GenBank mRNA | AK223234.1 | - | 3, 4 | 321 | ||
AK300837.1 | - | 3, 4 | 235 | |||
AK301888.1 | - | 4, 5 | 594 | |||
AK313255.1 | - | 3, 4 | 324 | |||
AK316406.1 | - | 4, 5 | 671 | |||
BC022015.2 | - | 3, 4 | 408 | 4824832 | ||
BC036307.1 | - | 3, 4 | 394 | 5165618 | ||
BG718473.1 | - | 3, 4 | 458 | |||
D17408.1 | - | 3, 4 | 336 | |||
D86058.1 | - | 2, 3 | 196 | |||
EU794601.1 | - | 3, 4 | 394 | |||
S80560.1 | - | 3, 4 | 318 | |||
U37019.1 | - | 3, 4 | 322 |