Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CALR SIRNA Assays ›
          Gene Symbol:
          CALR
          Gene Name:
          calreticulin
          Gene Aliases:
          CRT, HEL-S-99n, RO, SSA, cC1qR
          Chromosome Location:
          Chr.19: 12938600 - 12944490 on Build GRCh38
          UniGene ID:
          Hs.515162
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 14584

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CALR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004343.3 NP_004334.1 4, 5 580
          GenBank mRNA AB451408.1 - 4, 5 500
          AF087986.1 - 2, 3 292
          AK223060.1 - 4, 5 468
          AK295230.1 - 3, 4 523
          AK304492.1 - 3, 4 527
          AY047586.1 - 4, 5 553
          BC002500.2 - 4, 5 533 3050717
          BC007911.1 - 4, 5 570 4299303
          BC020493.1 - 4, 5 562 3898550
          BT007448.1 - 4, 5 500
          CR457070.1 - 4, 5 500
          FJ224311.1 - 4, 5 566
          M32294.1 - 4, 5 566
          M84739.1 - 4, 5 608

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          antigen processing and presentation of peptide antigen via MHC class I
          peptide antigen assembly with MHC class I protein complex
          regulation of transcription, DNA-templated
          protein folding
          protein export from nucleus
          cellular calcium ion homeostasis
          receptor-mediated endocytosis
          cell cycle arrest
          spermatogenesis
          positive regulation of cell proliferation
          positive regulation of gene expression
          negative regulation of translation
          protein maturation by protein folding
          cortical actin cytoskeleton organization
          response to estradiol
          negative regulation of intracellular steroid hormone receptor signaling pathway
          response to testosterone
          protein localization to nucleus
          protein folding in endoplasmic reticulum
          ATF6-mediated unfolded protein response
          regulation of meiotic nuclear division
          response to drug
          glucocorticoid receptor signaling pathway
          regulation of apoptotic process
          negative regulation of neuron differentiation
          positive regulation of DNA replication
          positive regulation of cell cycle
          negative regulation of transcription, DNA-templated
          negative regulation of retinoic acid receptor signaling pathway
          positive regulation of phagocytosis
          protein stabilization
          sequestering of calcium ion
          cardiac muscle cell differentiation
          chaperone-mediated protein folding
          cellular response to lithium ion
          cellular senescence
          positive regulation of substrate adhesion-dependent cell spreading
          positive regulation of dendritic cell chemotaxis
          complement component C1q binding
          glycoprotein binding
          DNA binding
          mRNA binding
          integrin binding
          iron ion binding
          calcium ion binding
          protein binding
          zinc ion binding
          carbohydrate binding
          ubiquitin protein ligase binding
          peptide binding
          hormone binding
          protein binding involved in protein folding
          poly(A) RNA binding
          androgen receptor binding
          unfolded protein binding
          chaperone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-49ts5:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0