Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EYA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282562.1 | NP_001269491.1 | 6 | 495 | ||
NM_001990.3 | NP_001981.2 | 7 | 575 | |||
NR_104214.1 | 7 | 575 | ||||
XM_006710449.3 | XP_006710512.1 | 8 | 611 | |||
XM_011540999.2 | XP_011539301.1 | 8 | 614 | |||
XM_011541001.2 | XP_011539303.1 | 7 | 572 | |||
XM_017000652.1 | XP_016856141.1 | 8 | 615 | |||
XM_017000653.1 | XP_016856142.1 | 9 | 735 | |||
XM_017000654.1 | XP_016856143.1 | 7 | 572 | |||
XM_017000655.1 | XP_016856144.1 | 8 | 728 | |||
XM_017000658.1 | XP_016856147.1 | 6 | 492 | |||
GenBank mRNA | AK289805.1 | - | 7 | 534 | ||
AK298129.1 | - | 6 | 492 | |||
BC029500.1 | - | 7 | 535 | 5272319 | ||
BX648945.1 | - | 7 | 544 | |||
U81602.1 | - | 2 | 174 |