Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BPTF SIRNA Assays ›
          Gene Symbol:
          BPTF AMZ2
          Gene Name:
          bromodomain PHD finger transcription factor
          archaelysin family metallopeptidase 2
          Gene Aliases:
          FAC1, FALZ, NURF301
          -
          Chromosome Location:
          Chr.17: 67825517 - 67984378 on Build GRCh38
          Chromosome Location:
          Chr.17: 68205486 - 68257164 on Build GRCh38
          UniGene ID:
          Hs.444200
          Hs.293560
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 14641

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BPTF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004459.6 NP_004450.3 1 653
          NM_182641.3 NP_872579.2 1 653
          XM_005257150.3 XP_005257207.1 1 800
          XM_005257151.3 XP_005257208.1 1 800
          XM_005257152.2 XP_005257209.1 1 800
          XM_005257153.3 XP_005257210.1 1 800
          XM_005257154.3 XP_005257211.1 1 800
          XM_005257155.3 XP_005257212.1 1 800
          XM_005257156.3 XP_005257213.1 1 800
          XM_005257157.3 XP_005257214.1 1 800
          XM_005257158.3 XP_005257215.1 1 800
          XM_005257159.2 XP_005257216.1 1 800
          XM_005257160.2 XP_005257217.1 1 800
          XM_005257161.3 XP_005257218.1 1 800
          XM_011524520.2 XP_011522822.1 1 800
          XM_011524521.2 XP_011522823.1 1 800
          XM_011524522.2 XP_011522824.1 1 800
          XM_011524523.2 XP_011522825.1 1 800
          XM_011524525.2 XP_011522827.1 1 800
          XM_011524526.2 XP_011522828.1 1 800
          XM_017024353.1 XP_016879842.1 1 800
          GenBank mRNA AB032251.1 - 1 646
          AK293224.1 - 1 227
          AK297281.1 - 2 320
          AY282495.1 - 1 175
          BC037422.1 - 1 804
          U05237.1 - 1 248
          Gene Symbol: AMZ2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_005257432.4 XP_005257489.1 1 732
          XM_017024723.1 XP_016880212.1 1 253
          XM_017024728.1 XP_016880217.1 1 163

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          embryonic placenta development
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          brain development
          endoderm development
          response to wounding
          anterior/posterior pattern specification
          covalent chromatin modification
          positive regulation of transcription, DNA-templated
          cellular response to nerve growth factor stimulus
          proteolysis
          protein binding
          DNA-dependent ATPase activity
          transcription factor binding
          zinc ion binding
          sequence-specific DNA binding
          metallopeptidase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0