Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165928.3 | NP_001159400.2 | 6 | 1362 | ||
NM_001177634.2 | NP_001171105.1 | 6 | 1305 | |||
NM_001177635.2 | NP_001171106.1 | 4 | 894 | |||
NM_001177636.2 | NP_001171107.1 | 5 | 1283 | |||
NM_001177637.2 | NP_001171108.1 | 4 | 1121 | |||
NM_001177638.2 | NP_001171109.1 | 4 | 1152 | |||
NM_001177639.2 | NP_001171110.1 | 3 | 842 | |||
NM_001177640.2 | NP_001171111.1 | 4 | 1058 | |||
NM_001177641.2 | NP_001171112.1 | 4 | 1148 | |||
NM_001177642.2 | NP_001171113.1 | 4 | 1053 | |||
NM_001177643.2 | NP_001171114.1 | 3 | 1052 | |||
NM_001177644.2 | NP_001171115.1 | 3 | 1141 | |||
NM_004393.5 | NP_004384.4 | 3 | 1001 | |||
GenBank mRNA | AK291692.1 | - | 3 | 843 | ||
AK297802.1 | - | 1 | 157 | |||
BC012740.2 | - | 3 | 936 | 4099382 | ||
BC014616.2 | - | 3 | 936 | 2964740 | ||
L19711.1 | - | 3 | 977 |