Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RCAN1 SIRNA Assays ›
          Gene Symbol:
          RCAN1
          Gene Name:
          regulator of calcineurin 1
          Gene Aliases:
          ADAPT78, CSP1, DSC1, DSCR1, MCIP1, RCN1
          Chromosome Location:
          Chr.21: 34516442 - 34615142 on Build GRCh38
          UniGene ID:
          Hs.282326
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146452

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RCAN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001285389.2 NP_001272318.1 2 127
          NM_001285391.2 NP_001272320.2 2 450
          NM_001285392.2 NP_001272321.1 2 244
          NM_001285393.2 NP_001272322.1 2 181
          NM_004414.6 NP_004405.3 2 453
          NM_203417.2 NP_981962.1 2 239
          NM_203418.2 NP_981963.1 2 418
          XM_005260929.1 XP_005260986.1 2 374
          XM_005260930.1 XP_005260987.1 2 362
          XM_017028282.1 XP_016883771.1 3 779
          XM_017028283.1 XP_016883772.1 3 779
          GenBank mRNA AF400429.1 - 2 111
          AK055845.1 - 1 650
          AK092184.1 - 2 374
          AK131569.1 - 2 371
          AK293293.1 - 2 232
          AK299444.1 - 2 168
          AL556803.3 - 2 453
          AY325903.1 - 2 445
          BC002864.2 - 2 213 3944959
          BT007363.1 - 2 157
          BX419260.2 - 2 371
          CR456878.1 - 1 79
          CR542065.1 - 2 157
          DA503835.1 - 3 334
          DA586027.1 - 2 181
          DB023862.1 - 2 244
          DC331441.1 - 2 418
          U28833.2 - 2 127
          U85265.2 - 2 238
          U85266.2 - 2 181
          U85267.2 - 2 300

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          signal transduction
          central nervous system development
          blood circulation
          calcineurin-NFAT signaling cascade
          regulation of catalytic activity
          regulation of calcineurin-NFAT signaling cascade
          nucleotide binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          calcium-dependent protein serine/threonine phosphatase regulator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0