Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: EPHB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004445.5 | NP_004436.4 | 3 | 306 | ||
NR_104001.2 | 4 | 423 | ||||
XM_006715881.2 | XP_006715944.1 | 6 | 1301 | |||
XM_011515879.1 | XP_011514181.1 | 7 | 1340 | |||
XM_011515880.1 | XP_011514182.1 | 6 | 1318 | |||
XM_011515881.2 | XP_011514183.1 | 6 | 1302 | |||
XM_011515882.2 | XP_011514184.1 | 6 | 1304 | |||
GenBank mRNA | BC110607.1 | - | 3 | 303 | 40028687 | |
BX410199.2 | - | 2 | 228 | |||
D83492.1 | - | 6 | 880 | |||
EU054308.1 | - | 2 | 82 |