Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other COL13A1 SIRNA Assays ›
Gene Symbol:
COL13A1
Gene Name:
collagen type XIII alpha 1 chain
Gene Aliases:
CMS19, COLXIIIA1
Chromosome Location:
Chr.10: 69801885 - 69959148 on Build GRCh38
UniGene ID:
Hs.695934
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146594
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: COL13A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130103.1
NP_001123575.1
1
815
NM_001320951.1
NP_001307880.1
1
815
NM_080798.3
NP_542988.3
1
815
NM_080800.3
NP_542990.3
1
815
NM_080801.3
NP_542991.3
1
815
NM_080802.3
NP_542992.3
1
815
NM_080805.3
NP_542995.3
1
815
XM_011539292.2
XP_011537594.1
1
818
XM_011539293.2
XP_011537595.1
1
818
XM_011539294.2
XP_011537596.1
1
818
XM_011539295.2
XP_011537597.1
1
818
XM_017015676.1
XP_016871165.1
1
818
XM_017015677.1
XP_016871166.1
1
818
XM_017015678.1
XP_016871167.1
1
818
XM_017015679.1
XP_016871168.1
1
818
XM_017015680.1
XP_016871169.1
1
818
XM_017015681.1
XP_016871170.1
1
818
XM_017015682.1
XP_016871171.1
1
818
XM_017015683.1
XP_016871172.1
1
818
XM_017015684.1
XP_016871173.1
1
818
XM_017015685.1
XP_016871174.1
1
818
XM_017015686.1
XP_016871175.1
1
818
XM_017015687.1
XP_016871176.1
1
818
XM_017015688.1
XP_016871177.1
1
818
XM_017015689.1
XP_016871178.1
1
818
XM_017015690.1
XP_016871179.1
1
818
XM_017015691.1
XP_016871180.1
1
818
XM_017015692.1
XP_016871181.1
1
818
XM_017015693.1
XP_016871182.1
1
818
XM_017015694.1
XP_016871183.1
1
818
XM_017015695.1
XP_016871184.1
1
818
XM_017015697.1
XP_016871186.1
1
196
GenBank mRNA
AJ293624.1
-
1
776
BC136385.1
-
1
585
9020372
BC139913.1
-
1
787
M59217.1
-
1
151
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix structural protein
Gene Ontology Categories:
Function(s)
Process(es)
morphogenesis of a branching structure
endochondral ossification
cell-matrix adhesion
single organismal cell-cell adhesion
cell differentiation
extracellular matrix organization
collagen catabolic process
protein binding
heparin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0