Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DDX11 SIRNA Assays ›
          Gene Symbol:
          DDX11 DDX12P LOC642846
          Gene Name:
          DEAD/H-box helicase 11
          DEAD/H-box helicase 12, pseudogene
          DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11-like
          Gene Aliases:
          CHL1, CHLR1, KRG2, WABS
          CHLR2, DDX12
          -
          Chromosome Location:
          Chr.12: 31073574 - 31104799 on Build GRCh38
          Chromosome Location:
          Chr.12: 9417691 - 9448172 on Build GRCh38
          Chromosome Location:
          Chr.12: 9283657 - 9314088 on Build GRCh38
          UniGene ID:
          Hs.443960
          Hs.447869
          Hs.447869
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID 146796

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DDX11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257144.1 NP_001244073.1 27 3267
          NM_001257145.1 NP_001244074.1 27 3199
          NM_004399.2 NP_004390.3 26 3244
          NM_030653.3 NP_085911.2 27 3394
          NM_152438.1 NP_689651.1 27 3389
          XM_005253331.3 XP_005253388.1 27 3559
          XM_005253333.3 XP_005253390.1 27 3564
          XM_006719050.3 XP_006719113.1 26 3639
          XM_011520592.2 XP_011518894.1 26 3635
          XM_011520595.2 XP_011518897.1 26 3414
          XM_017018918.1 XP_016874407.1 27 3817
          XM_017018919.1 XP_016874408.1 28 3880
          XM_017018920.1 XP_016874409.1 27 3779
          XM_017018921.1 XP_016874410.1 28 3742
          XM_017018922.1 XP_016874411.1 27 3610
          XM_017018923.1 XP_016874412.1 27 3503
          XM_017018924.1 XP_016874413.1 27 3536
          XM_017018925.1 XP_016874414.1 27 3608
          XM_017018926.1 XP_016874415.1 27 3677
          XM_017018927.1 XP_016874416.1 26 3668
          XM_017018928.1 XP_016874417.1 27 3822
          XM_017018929.1 XP_016874418.1 27 3784
          XM_017018930.1 XP_016874419.1 28 3746
          XM_017018931.1 XP_016874420.1 26 3435
          XM_017018932.1 XP_016874421.1 27 3253
          XM_017018933.1 XP_016874422.1 27 3904
          XM_017018934.1 XP_016874423.1 25 3356
          XM_017018935.1 XP_016874424.1 23 2840
          XM_017018936.1 XP_016874425.1 23 3835
          XM_017018937.1 XP_016874426.1 23 3840
          GenBank mRNA BC012834.2 - 3 651
          BC050069.1 - 27 3184 5271949
          BC050522.1 - 27 3218 6471750
          DQ786287.1 - 1 300
          U75967.1 - 27 3284
          U75968.1 - 27 3649
          U75969.1 - 19 2239
          X99583.1 - 26 3205
          Gene Symbol: DDX12P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_033399.1 27 3352
          GenBank mRNA BC065494.1 - 15 3366
          DQ786301.1 - 1 259
          Gene Symbol: LOC642846
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024374.1 22 2964
          GenBank mRNA AW391236.1 - 1 289

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          sister chromatid cohesion
          viral process
          DNA duplex unwinding
          IRE1-mediated unfolded protein response
          cell cycle
          double-stranded DNA binding
          single-stranded DNA binding
          RNA binding
          ATP-dependent DNA helicase activity
          helicase activity
          protein binding
          ATP binding
          DNA-dependent ATPase activity
          metal ion binding
          4 iron, 4 sulfur cluster binding
          DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-bqxw8:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0