Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAMK2G SIRNA Assays ›
Gene Symbol:
CAMK2G
Gene Name:
calcium/calmodulin dependent protein kinase II gamma
Gene Aliases:
CAMK, CAMK-II, CAMKG
Chromosome Location:
Chr.10: 73812501 - 73874591 on Build GRCh38
UniGene ID:
Hs.523045
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146841
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAMK2G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204492.1
NP_001191421.1
3
303
NM_001222.3
NP_001213.2
3
303
NM_001320898.1
NP_001307827.1
3
303
NM_172169.2
NP_751909.1
3
303
NM_172170.4
NP_751910.1
3
303
NM_172171.2
NP_751911.1
3
303
NM_172173.2
NP_751913.1
3
303
XM_005270195.1
XP_005270252.1
3
294
XM_005270197.1
XP_005270254.1
3
294
XM_005270198.1
XP_005270255.1
3
294
XM_005270199.1
XP_005270256.1
3
294
XM_005270200.1
XP_005270257.1
3
294
XM_005270201.1
XP_005270258.1
3
294
XM_005270203.1
XP_005270260.1
3
294
XM_005270205.1
XP_005270262.1
3
294
XM_006717993.1
XP_006718056.1
3
294
XM_006717996.1
XP_006718059.1
3
294
XM_006717997.1
XP_006718060.1
3
294
XM_006717998.1
XP_006718061.1
3
294
XM_011540225.2
XP_011538527.1
3
242
XM_017016716.1
XP_016872205.1
3
294
XM_017016717.1
XP_016872206.1
3
242
XM_017016718.1
XP_016872207.1
3
294
XM_017016719.1
XP_016872208.1
3
294
XM_017016720.1
XP_016872209.1
3
294
XM_017016721.1
XP_016872210.1
3
369
XM_017016722.1
XP_016872211.1
3
294
XM_017016723.1
XP_016872212.1
3
294
XM_017016724.1
XP_016872213.1
3
294
XM_017016725.1
XP_016872214.1
3
294
XM_017016726.1
XP_016872215.1
3
294
XM_017016727.1
XP_016872216.1
3
294
XM_017016728.1
XP_016872217.1
3
242
XM_017016729.1
XP_016872218.1
2
84
XM_017016730.1
XP_016872219.1
3
294
XM_017016731.1
XP_016872220.1
3
255
XM_017016732.1
XP_016872221.1
3
188
XM_017016733.1
XP_016872222.1
3
242
XM_017016734.1
XP_016872223.1
3
281
XM_017016735.1
XP_016872224.1
3
294
XM_017016736.1
XP_016872225.1
3
369
XM_017016737.1
XP_016872226.1
3
251
XM_017016738.1
XP_016872227.1
3
255
XM_017016739.1
XP_016872228.1
3
426
GenBank mRNA
AK095576.1
-
4
400
AK131199.1
-
3
226
AK225771.1
-
3
303
AK291704.1
-
3
295
AK301180.1
-
3
295
BC034044.1
-
3
294
4813948
BU165166.1
-
3
300
DR002605.1
-
3
310
L07044.1
-
3
194
U66064.1
-
3
226
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
MAPK cascade
protein dephosphorylation
calcium ion transport
nervous system development
regulation of skeletal muscle adaptation
insulin secretion
cell differentiation
positive regulation of GTPase activity
protein autophosphorylation
protein oligomerization
regulation of calcium ion transport
interferon-gamma-mediated signaling pathway
regulation of cellular response to heat
regulation of relaxation of cardiac muscle
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
calcium-dependent protein serine/threonine phosphatase activity
Ras guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-6tb5w:80/100.66.72.8:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0