Search
Search

ACVR1| Gene Symbol: ACVR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105.4 | NP_001096.1 | 5 | 921 | ||
| NM_001111067.2 | NP_001104537.1 | 5 | 740 | |||
| XM_005246939.3 | XP_005246996.1 | 5 | 792 | |||
| XM_005246940.3 | XP_005246997.1 | 5 | 760 | |||
| XM_006712825.3 | XP_006712888.1 | 5 | 873 | |||
| XM_011512106.2 | XP_011510408.1 | 6 | 840 | |||
| XM_011512107.2 | XP_011510409.1 | 5 | 812 | |||
| XM_011512108.2 | XP_011510410.1 | 5 | 2219 | |||
| GenBank mRNA | AK297765.1 | - | 5 | 708 | ||
| AK313630.1 | - | 5 | 716 | |||
| BC033867.1 | - | 5 | 921 | 5209711 | ||
| BC036748.1 | - | 5 | 921 | |||
| KU177885.1 | - | 3 | 491 | |||
| KU177886.1 | - | 3 | 491 | |||
| KU177887.1 | - | 3 | 491 | |||
| L02911.1 | - | 6 | 831 | |||
| Z22534.1 | - | 4 | 594 | |||