Search
Search

ANPEP| Gene Symbol: ANPEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001150.2 | NP_001141.2 | 21 | 3136 | ||
| XM_005254892.4 | XP_005254949.1 | 21 | 3144 | |||
| XM_011521473.1 | XP_011519775.1 | 21 | 3287 | |||
| GenBank mRNA | AB209918.1 | - | 21 | 3132 | ||
| AK298134.1 | - | 15 | 1606 | |||
| AK298242.1 | - | 15 | 1657 | |||
| AK298339.1 | - | 16 | 1717 | |||
| AK300946.1 | - | 17 | 1867 | |||
| BC058928.1 | - | 20 | 2937 | 6020902 | ||
| M22324.1 | - | 20 | 2964 | |||
| X13276.1 | - | 20 | 2868 | |||