Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANXA8 SIRNA Assays ›
          Gene Symbol:
          ANXA8 ANXA8L1
          Gene Name:
          annexin A8
          annexin A8-like 1
          Gene Aliases:
          ANX8, CH17-360D5.2
          ANXA8, ANXA8L2, VAC-beta, bA145E20.2
          Chromosome Location:
          Chr.10: 47467993 - 47991879 on Build GRCh38
          Chromosome Location:
          Chr.10: 46375590 - 46391784 on Build GRCh38
          UniGene ID:
          Hs.535306
          Hs.705389
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 147157

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANXA8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001040084.2 NP_001035173.1 3 349
          NM_001271702.1 NP_001258631.1 3 463
          XM_006717951.3 XP_006718014.1 3 303
          XM_011540096.1 XP_011538398.1 4 502
          XM_011540098.1 XP_011538400.1 4 502
          XM_011540099.1 XP_011538401.1 4 502
          XM_011540101.2 XP_011538403.1 4 502
          GenBank mRNA AK296976.1 - 3 253
          AK298761.1 - 3 329
          AK300139.1 - 3 314
          AK300158.1 - 3 207
          BC004376.1 - 3 246 3639064
          BC073755.1 - 3 280 6538697
          CB270161.1 - 3 335
          DC339657.1 - 3 349
          M81844.1 - 3 151
          Z36830.1 - 3 260
          Gene Symbol: ANXA8L1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098845.2 NP_001092315.2 3 386
          NM_001278923.1 NP_001265852.1 3 386
          NM_001278924.1 NP_001265853.1 3 500
          GenBank mRNA AK223149.1 - 3 200
          AK300191.1 - 3 220
          AK307182.1 - 3 200
          AK313626.1 - 3 249
          BC008813.2 - 3 414 3958020
          DC381265.1 - 3 387
          X16662.1 - 3 230

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          calcium-binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endosome organization
          blood coagulation
          endosomal transport
          negative regulation of serine-type endopeptidase activity
          negative regulation of phospholipase A2 activity
          calcium ion binding
          calcium-dependent phospholipid binding
          phosphatidylinositol-4,5-bisphosphate binding
          phosphatidylinositol-3,4,5-trisphosphate binding
          phosphatidylinositol-3,4-bisphosphate binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ptcs5:80/100.66.79.248:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0