Search
Search

BTC| Gene Symbol: BTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001729.3 | NP_001720.1 | 4 | 673 | ||
| XM_011532211.1 | XP_011530513.1 | 4 | 654 | |||
| GenBank mRNA | BC011618.2 | - | 4 | 464 | 3952444 | |
| KU177946.1 | - | 4 | 312 | |||
| S55606.1 | - | 4 | 606 | |||