Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZC3H3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015117.2 | NP_055932.2 | 2 | 403 | ||
XM_006716536.3 | XP_006716599.2 | 4 | 792 | |||
XM_011516943.2 | XP_011515245.1 | 2 | 518 | |||
XM_011516944.2 | XP_011515246.1 | 2 | 518 | |||
XM_017013248.1 | XP_016868737.1 | 3 | 1423 | |||
XM_017013249.1 | XP_016868738.1 | 3 | 965 | |||
GenBank mRNA | BC038670.1 | - | 2 | 386 | 5260824 | |
BM148116.1 | - | 2 | 406 | |||
D63484.1 | - | 2 | 363 |