Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TEX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161584.1 | NP_001155056.1 | 5 | 1346 | ||
NM_017746.3 | NP_060216.2 | 5 | 1402 | |||
XM_011518798.1 | XP_011517100.1 | 5 | 1364 | |||
XM_017014848.1 | XP_016870337.1 | 5 | 891 | |||
GenBank mRNA | AB060968.1 | - | 5 | 1318 | ||
AK000294.1 | - | 5 | 1318 | |||
AK298916.1 | - | 7 | 925 | |||
AK302619.1 | - | 5 | 1349 | |||
AK303662.1 | - | 5 | 842 | |||
AK304869.1 | - | 4 | 1119 | |||
AY598337.1 | - | 4 | 1187 | |||
BC030652.2 | - | 5 | 1238 | 5268105 |