Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCDC7 SIRNA Assays ›
          Gene Symbol:
          CCDC7
          Gene Name:
          coiled-coil domain containing 7
          Gene Aliases:
          BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
          Chromosome Location:
          Chr.10: 32446082 - 32882864 on Build GRCh38
          UniGene ID:
          Hs.585464
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 147865

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCDC7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321115.1 NP_001308044.1 26, 27 3163
          XM_005252593.2 XP_005252650.1 7, 8 950
          XM_006717505.1 XP_006717568.1 24, 25 2658
          XM_006717506.1 XP_006717569.1 23, 24 2550
          XM_011519663.1 XP_011517965.1 26, 27 2871
          XM_011519664.1 XP_011517966.1 26, 27 2871
          XM_011519666.1 XP_011517968.1 26, 27 2871
          XM_011519667.1 XP_011517969.1 26, 27 2871
          XM_011519668.1 XP_011517970.1 25, 26 2781
          XM_011519669.1 XP_011517971.1 25, 26 2769
          XM_011519670.1 XP_011517972.1 25, 26 2763
          XM_011519671.1 XP_011517973.1 26, 27 2871
          XM_011519672.1 XP_011517974.1 25, 26 2760
          XM_011519673.1 XP_011517975.1 25, 26 2757
          XM_011519674.1 XP_011517976.1 26, 27 2871
          XM_011519675.1 XP_011517977.1 24, 25 2661
          XM_011519676.1 XP_011517978.1 25, 26 2757
          XM_011519677.1 XP_011517979.1 25, 26 2649
          XM_011519678.1 XP_011517980.1 24, 25 2646
          XM_011519679.1 XP_011517981.1 23, 24 2340
          XM_011519680.2 XP_011517982.1 21, 22 2006
          XM_011519683.2 XP_011517985.1 15, 16 1680
          XM_011519684.2 XP_011517986.1 26, 27 2871
          XM_011519685.1 XP_011517987.1 18, 19 1888
          XM_011519686.2 XP_011517988.1 26, 27 2871
          XM_011519687.1 XP_011517989.1 8, 9 795
          XM_017016638.1 XP_016872127.1 26, 27 2871
          XM_017016640.1 XP_016872129.1 26, 27 2871
          XM_017016641.1 XP_016872130.1 21, 22 2300
          XM_017016642.1 XP_016872131.1 21, 22 2373
          XM_017016643.1 XP_016872132.1 21, 22 1990
          XM_017016644.1 XP_016872133.1 26, 27 2871
          XM_017016645.1 XP_016872134.1 20, 21 1914
          XM_017016646.1 XP_016872135.1 17, 18 1801
          XM_017016647.1 XP_016872136.1 26, 27 2871
          XM_017016648.1 XP_016872137.1 13, 14 1522
          XM_017016649.1 XP_016872138.1 13, 14 1462
          XM_017016652.1 XP_016872141.1 10, 11 925
          XM_017016653.1 XP_016872142.1 8, 9 835
          GenBank mRNA AK023093.1 - 7, 8 956
          AK301824.1 - 4, 5 391
          BC125090.1 - 10, 11 1125 40118176
          BC125091.1 - 5, 6 481 40118177
          DB026046.1 - 3, 4 309

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lmjtd:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0