Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRDMT1 SIRNA Assays ›
          Gene Symbol:
          TRDMT1
          Gene Name:
          tRNA aspartic acid methyltransferase 1
          Gene Aliases:
          DMNT2, DNMT2, MHSAIIP, PUMET, RNMT1
          Chromosome Location:
          Chr.10: 17136785 - 17202071 on Build GRCh38
          UniGene ID:
          Hs.351665
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 14806

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRDMT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321006.1 NP_001307935.1 7, 8 1247
          NM_001321007.1 NP_001307936.1 10, 11 1620
          NM_004412.6 NP_004403.1 9, 10 1390
          XM_005252372.4 XP_005252429.1 9, 10 1385
          XM_005252374.4 XP_005252431.1 9, 10 1385
          XM_005252375.4 XP_005252432.1 7, 8 1247
          XM_005252378.3 XP_005252435.1 10, 11 1216
          XM_006717388.3 XP_006717451.1 9, 10 1364
          XM_011519343.2 XP_011517645.1 8, 9 1319
          XM_011519344.2 XP_011517646.1 8, 9 1308
          XM_011519345.2 XP_011517647.1 8, 9 1721
          XM_017015800.1 XP_016871289.1 7, 8 1226
          XM_017015801.1 XP_016871290.1 7, 8 1247
          XM_017015802.1 XP_016871291.1 9, 10 3604
          XM_017015803.1 XP_016871292.1 10, 11 1218
          XM_017015804.1 XP_016871293.1 10, 11 1056
          XM_017015805.1 XP_016871294.1 9, 10 1253
          XM_017015806.1 XP_016871295.1 10, 11 1195
          XM_017015807.1 XP_016871296.1 10, 11 1216
          XM_017015808.1 XP_016871297.1 10, 11 1030
          XM_017015809.1 XP_016871298.1 6, 7 1170
          XM_017015810.1 XP_016871299.1 9, 10 1150
          XM_017015811.1 XP_016871300.1 7, 8 1116
          XM_017015812.1 XP_016871301.1 7, 8 910
          GenBank mRNA AF012128.1 - 9, 10 960
          AF045888.1 - 9, 10 959
          AJ223333.1 - 9, 10 1001
          AK291417.1 - 9, 10 959
          AK294100.1 - 8, 9 889
          AK299027.1 - 7, 8 1027
          AK303428.1 - 4, 5 607
          BC047733.1 - 9, 10 960 6473993
          BX537961.1 - 10, 11 1210
          CR450316.1 - 9, 10 953
          HQ637546.1 - 7, 8 788
          HQ637547.1 - 8, 9 855
          HQ637548.1 - 7, 8 810
          HQ637549.1 - 8, 9 887

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to amphetamine
          tRNA modification
          tRNA methylation
          C-5 methylation of cytosine
          RNA binding
          DNA (cytosine-5-)-methyltransferase activity
          tRNA methyltransferase activity
          tRNA (cytosine-5-)-methyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-s97cd:80/100.66.77.220:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0