Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC7 SIRNA Assays ›
Gene Symbol:
CCDC7
Gene Name:
coiled-coil domain containing 7
Gene Aliases:
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location:
Chr.10: 32446082 - 32882864 on Build GRCh38
UniGene ID:
Hs.585464
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
148069
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001026383.2
NP_001021554.1
7
1013
NM_001321115.1
NP_001308044.1
7
1247
NM_145023.5
NP_659460.3
7
1247
NR_109826.1
7
1013
XM_006717505.1
XP_006717568.1
7
955
XM_006717506.1
XP_006717569.1
7
955
XM_011519663.1
XP_011517965.1
7
955
XM_011519664.1
XP_011517966.1
7
955
XM_011519666.1
XP_011517968.1
7
955
XM_011519667.1
XP_011517969.1
7
955
XM_011519668.1
XP_011517970.1
7
955
XM_011519669.1
XP_011517971.1
7
955
XM_011519670.1
XP_011517972.1
7
955
XM_011519671.1
XP_011517973.1
7
955
XM_011519672.1
XP_011517974.1
7
955
XM_011519673.1
XP_011517975.1
7
955
XM_011519674.1
XP_011517976.1
7
955
XM_011519675.1
XP_011517977.1
7
955
XM_011519676.1
XP_011517978.1
7
955
XM_011519677.1
XP_011517979.1
7
955
XM_011519678.1
XP_011517980.1
7
955
XM_011519679.1
XP_011517981.1
7
955
XM_011519680.2
XP_011517982.1
2
90
XM_011519684.2
XP_011517986.1
7
955
XM_011519685.1
XP_011517987.1
1
93
XM_011519686.2
XP_011517988.1
7
955
XM_011519688.2
XP_011517990.1
7
955
XM_017016638.1
XP_016872127.1
7
955
XM_017016639.1
XP_016872128.1
7
955
XM_017016640.1
XP_016872129.1
7
955
XM_017016641.1
XP_016872130.1
2
384
XM_017016642.1
XP_016872131.1
2
457
XM_017016644.1
XP_016872133.1
7
955
XM_017016647.1
XP_016872136.1
7
955
XM_017016650.1
XP_016872139.1
7
955
XM_017016651.1
XP_016872140.1
7
955
GenBank mRNA
AK057324.1
-
7
955
AK058148.1
-
5
359
BC022020.2
-
7
1153
4825109
BC042672.1
-
7
938
5166976
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-26xzd:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0