Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCDC7 SIRNA Assays ›
          Gene Symbol:
          CCDC7
          Gene Name:
          coiled-coil domain containing 7
          Gene Aliases:
          BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
          Chromosome Location:
          Chr.10: 32446082 - 32882864 on Build GRCh38
          UniGene ID:
          Hs.585464
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 148071

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCDC7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001026383.2 NP_001021554.1 16 1585
          NM_001321115.1 NP_001308044.1 16 1819
          NM_145023.5 NP_659460.3 16 1819
          NR_109826.1 15 1509
          XM_006717505.1 XP_006717568.1 16 1527
          XM_006717506.1 XP_006717569.1 16 1527
          XM_011519663.1 XP_011517965.1 16 1527
          XM_011519664.1 XP_011517966.1 16 1527
          XM_011519666.1 XP_011517968.1 16 1527
          XM_011519667.1 XP_011517969.1 16 1527
          XM_011519668.1 XP_011517970.1 15 1437
          XM_011519669.1 XP_011517971.1 16 1527
          XM_011519670.1 XP_011517972.1 16 1527
          XM_011519671.1 XP_011517973.1 16 1527
          XM_011519672.1 XP_011517974.1 16 1527
          XM_011519673.1 XP_011517975.1 16 1527
          XM_011519674.1 XP_011517976.1 16 1527
          XM_011519675.1 XP_011517977.1 16 1527
          XM_011519676.1 XP_011517978.1 16 1527
          XM_011519678.1 XP_011517980.1 16 1527
          XM_011519680.2 XP_011517982.1 11 662
          XM_011519683.2 XP_011517985.1 5 336
          XM_011519684.2 XP_011517986.1 16 1527
          XM_011519685.1 XP_011517987.1 8 544
          XM_011519686.2 XP_011517988.1 16 1527
          XM_011519688.2 XP_011517990.1 16 1527
          XM_017016638.1 XP_016872127.1 16 1527
          XM_017016639.1 XP_016872128.1 16 1527
          XM_017016640.1 XP_016872129.1 16 1527
          XM_017016641.1 XP_016872130.1 11 956
          XM_017016642.1 XP_016872131.1 11 1029
          XM_017016643.1 XP_016872132.1 11 646
          XM_017016644.1 XP_016872133.1 16 1527
          XM_017016645.1 XP_016872134.1 10 570
          XM_017016646.1 XP_016872135.1 7 457
          XM_017016647.1 XP_016872136.1 16 1527
          XM_017016648.1 XP_016872137.1 3 178
          XM_017016649.1 XP_016872138.1 3 118
          XM_017016650.1 XP_016872139.1 16 1527
          XM_017016651.1 XP_016872140.1 16 1527
          GenBank mRNA AK057324.1 - 15 1451
          AK058148.1 - 14 931
          BC022020.2 - 16 1725 4825109
          BC042672.1 - 16 1510 5166976

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0