Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RASGEF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282862.1 | NP_001269791.1 | 10 | 1266 | ||
NM_145313.3 | NP_660356.2 | 10 | 1238 | |||
XM_005271809.3 | XP_005271866.1 | 9 | 1116 | |||
XM_011539500.2 | XP_011537802.1 | 9 | 1226 | |||
GenBank mRNA | AK095136.1 | - | 10 | 1266 | ||
AK127432.1 | - | 10 | 3664 | |||
BC022548.1 | - | 10 | 1238 | 4814750 |